Assay Details
| Gene Symbol: RBL12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101718.5 | NP_564058.1 | 6 - 7 | 1237 | 116 | |
| GenBank mRNA | AF370504.1 | - | 6 - 7 | 1041 | 116 | |
| AY081685.1 | - | 6 - 7 | 872 | 116 | ||
| AY086276.1 | - | 6 - 7 | 890 | 116 | ||
More Information
Set Membership: |
|

RBL12
Genomic Map