Assay Details
| Gene Symbol: GR-RBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101721.4 | NP_173298.1 | 2 - 3 | 349 | 92 | |
| GenBank mRNA | AK227482.1 | - | 2 - 3 | 235 | 92 | |
| BT005999.1 | - | 1 - 2 | 126 | 92 | ||
More Information
Set Membership: |
|

GR-RBP6
Genomic Map