Assay Details
| Gene Symbol: GR-RBP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101721.4 | NP_173298.1 | 3 - 4 | 417 | 71 | |
| GenBank mRNA | AK227482.1 | - | 3 - 4 | 303 | 71 | |
| BT005999.1 | - | 2 - 3 | 194 | 71 | ||
More Information
Set Membership: |
|

GR-RBP6
Genomic Map