Assay Details
| Gene Symbol: ICS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160879.1 | NP_001154351.1 | 1 - 2 | 263 | 74 | |
| NM_001332377.1 | NP_001321112.1 | 2 - 3 | 424 | 74 | ||
| NM_101744.7 | NP_173321.4 | 2 - 3 | 424 | 74 | ||
| GenBank mRNA | EU589462.1 | - | 2 - 3 | 368 | 74 | |
More Information
Set Membership: |
|

ICS2
Genomic Map