Assay Details
| Gene Symbol: ICS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160879.1 | NP_001154351.1 | 7 - 8 | 1026 | 72 | |
| NM_001332377.1 | NP_001321112.1 | 8 - 9 | 1187 | 72 | ||
| NM_101744.7 | NP_173321.4 | 8 - 9 | 1187 | 72 | ||
| GenBank mRNA | EU589462.1 | - | 8 - 9 | 1131 | 72 | |
More Information
Set Membership: |
|

ICS2
Genomic Map