Assay Details
Gene Symbol: ARR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101763.3 | NP_173339.1 | 4 - 5 | 616 | 107 | |
GenBank mRNA | AB008490.1 | - | 4 - 5 | 413 | 107 | |
BT024585.1 | - | 4 - 5 | 380 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
