Assay Details
| Gene Symbol: LSM1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101770.2 | NP_564072.1 | 4 - 5 | 484 | 71 | |
| GenBank mRNA | AF411792.1 | - | 4 - 5 | 455 | 71 | |
| AY094024.1 | - | 4 - 5 | 311 | 71 | ||
More Information
Set Membership: |
|

LSM1A
Genomic Map