Assay Details
| Gene Symbol: AT1G19490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101806.3 | NP_173381.1 | 4 - 5 | 714 | 141 | |
| GenBank mRNA | AY099875.1 | - | 4 - 5 | 696 | 141 | |
| BT000319.1 | - | 4 - 5 | 600 | 141 | ||
More Information
Set Membership: |
|

AT1G19490
Genomic Map