Assay Details
Gene Symbol: FNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084097.1 | NP_001077566.1 | 8 - 9 | 1178 | 101 | |
NM_001332422.1 | NP_001319047.1 | 7 - 8 | 1513 | 101 | ||
NM_101857.6 | NP_173431.1 | 8 - 9 | 1270 | 101 | ||
GenBank mRNA | AK318722.1 | - | 7 - 8 | 1400 | 101 | |
AY062739.1 | - | 8 - 9 | 1062 | 101 | ||
AY114663.1 | - | 8 - 9 | 1044 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
