Assay Details
| Gene Symbol: HYD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101859.4 | NP_173433.1 | 3 - 4 | 515 | 77 | |
| GenBank mRNA | AF030357.1 | - | 3 - 4 | 451 | 77 | |
| AF334733.1 | - | 3 - 4 | 398 | 77 | ||
| AK228406.1 | - | 3 - 4 | 461 | 77 | ||
More Information
Set Membership: |
|

HYD1
Genomic Map