Assay Details
Gene Symbol: SK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101868.5 | NP_564105.1 | 1 - 2 | 434 | 72 | |
GenBank mRNA | AY058205.1 | - | 1 - 2 | 385 | 72 | |
AY098980.1 | - | 1 - 2 | 316 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
