Assay Details
| Gene Symbol: TIM17-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101886.4 | NP_173460.1 | 1 | 617 | 75 | |
| GenBank mRNA | AK118449.1 | - | 1 | 539 | 75 | |
| AY463969.1 | - | 1 | 508 | 75 | ||
| BT005239.1 | - | 1 | 508 | 75 | ||
More Information
Set Membership: |
|

TIM17-1
Genomic Map