Assay Details
Gene Symbol: MCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101936.3 | NP_173507.1 | 1 - 2 | 87 | 88 | |
GenBank mRNA | AK118972.1 | - | 1 - 2 | 63 | 88 | |
BT006034.1 | - | 1 - 2 | 50 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
