Assay Details
Gene Symbol: MCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332470.1 | NP_001321244.1 | 3 - 4 | 914 | 112 | |
NM_101936.3 | NP_173507.1 | 4 - 5 | 928 | 112 | ||
GenBank mRNA | AK118972.1 | - | 4 - 5 | 904 | 112 | |
BT006034.1 | - | 4 - 5 | 891 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
