Assay Details
Gene Symbol: ESC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101943.2 | NP_173514.1 | 1 | 1370 | 123 | |
GenBank mRNA | AF194974.1 | - | 1 | 1176 | 123 | |
AK228296.1 | - | 1 | 1357 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
