Assay Details
| Gene Symbol: LSM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101972.5 | NP_173542.1 | 2 - 3 | 315 | 87 | |
| GenBank mRNA | AF334722.1 | - | 2 - 3 | 214 | 87 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LSM3A| Gene Symbol: LSM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101972.5 | NP_173542.1 | 2 - 3 | 315 | 87 | |
| GenBank mRNA | AF334722.1 | - | 2 - 3 | 214 | 87 | |
Set Membership: |
|