Assay Details
| Gene Symbol: GLTP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332497.1 | NP_001320933.1 | 4 - 5 | 964 | 104 | |
| NM_001332498.1 | NP_001320934.1 | 4 - 5 | 982 | 104 | ||
| NM_101988.5 | NP_173558.4 | 5 - 6 | 818 | 104 | ||
| GenBank mRNA | AK221557.1 | - | 5 - 6 | 750 | 104 | |
More Information
Set Membership: |
|

GLTP2
Genomic Map