Assay Details
| Gene Symbol: EMB1144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036080.2 | NP_001031157.1 | 1 - 2 | 274 | 105 | |
| NM_001036081.2 | NP_001031158.1 | 1 - 2 | 274 | 105 | ||
| NM_103779.5 | NP_564534.1 | 1 - 2 | 338 | 105 | ||
| GenBank mRNA | AK227099.1 | - | 1 - 2 | 208 | 105 | |
| AY057519.1 | - | 1 - 2 | 262 | 105 | ||
| BT015785.1 | - | 1 - 2 | 140 | 105 | ||
More Information
Set Membership: |
|

EMB1144
Genomic Map