Assay Details
Gene Symbol: ATRAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345748.1 | NP_001330043.1 | 4 - 5 | 652 | 83 | |
NM_126011.2 | NP_201414.1 | 4 - 5 | 652 | 83 | ||
GenBank mRNA | AB030250.1 | - | 4 - 5 | 552 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
