Assay Details
Gene Symbol: ATRAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126011.2 | NP_201414.1 | 6 - 7 | 1124 | 151 | |
GenBank mRNA | AB030250.1 | - | 6 - 7 | 1024 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
