Assay Details
| Gene Symbol: ATRAD17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345748.1 | NP_001330043.1 | 9 - 10 | 1764 | 84 | |
| NM_126011.2 | NP_201414.1 | 9 - 10 | 1812 | 84 | ||
| GenBank mRNA | AB030250.1 | - | 9 - 10 | 1712 | 84 | |
More Information
Set Membership: |
|

ATRAD17
Genomic Map