Assay Details
| Gene Symbol: CHR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126073.3 | NP_201476.1 | 14 - 15 | 2119 | 83 | |
| GenBank mRNA | AY099638.1 | - | 14 - 15 | 2065 | 83 | |
| BT002161.1 | - | 14 - 15 | 1966 | 83 | ||
More Information
Set Membership: |
|

CHR1
Genomic Map