Assay Details
Gene Symbol: SNRK2.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345807.1 | NP_001318893.1 | 7 - 8 | 901 | 72 | |
NM_126087.5 | NP_201489.1 | 6 - 7 | 819 | 72 | ||
GenBank mRNA | L05562.1 | - | 6 - 7 | 721 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
