Assay Details
| Gene Symbol: BT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001345832.1 | NP_001330470.1 | 3 - 4 | 630 | 100 | |
| NM_126148.4 | NP_201549.1 | 3 - 4 | 876 | 100 | ||
| NM_203278.2 | NP_975007.1 | 2 - 3 | 694 | 100 | ||
| GenBank mRNA | AF446873.1 | - | 2 - 3 | 365 | 100 | |
| AY052213.1 | - | 3 - 4 | 514 | 100 | ||
| AY316677.1 | - | 2 - 3 | 365 | 100 | ||
More Information
Set Membership: |
|

BT4
Genomic Map