Assay Details
| Gene Symbol: QSOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335047.1 | NP_001324094.1 | 8 - 9 | 1306 | 62 | |
| NM_001335048.1 | NP_001324093.1 | 9 - 10 | 1189 | 62 | ||
| NM_126188.4 | NP_565258.1 | 9 - 10 | 1041 | 62 | ||
| GenBank mRNA | AY090364.1 | - | 9 - 10 | 1015 | 62 | |
| AY125539.1 | - | 9 - 10 | 893 | 62 | ||
| BT000880.1 | - | 9 - 10 | 893 | 62 | ||
More Information
Set Membership: |
|

QSOX2
Genomic Map