Assay Details
| Gene Symbol: AT2G01440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126205.4 | NP_178253.3 | 7 - 8 | 1464 | 106 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G01440| Gene Symbol: AT2G01440 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126205.4 | NP_178253.3 | 7 - 8 | 1464 | 106 | |
Set Membership: |
|