Assay Details
Gene Symbol: WOL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335080.1 | NP_001324428.1 | 4 - 5 | 2094 | 72 | |
NM_001335081.1 | NP_001324427.1 | 5 - 6 | 1571 | 72 | ||
NM_001335082.1 | NP_001324429.1 | 5 - 6 | 1280 | 72 | ||
NM_126244.3 | NP_565277.1 | 5 - 6 | 1597 | 72 | ||
NM_179594.2 | NP_849925.1 | 5 - 6 | 1576 | 72 | ||
NM_201667.1 | NP_973396.1 | 6 - 7 | 1297 | 72 | ||
GenBank mRNA | AB046871.1 | - | 5 - 6 | 1235 | 72 | |
AJ278528.1 | - | 5 - 6 | 1305 | 72 | ||
AJ278529.1 | - | 5 - 6 | 1188 | 72 | ||
AJ278530.1 | - | 6 - 7 | 1297 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
