Assay Details
Gene Symbol: WOL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335080.1 | NP_001324428.1 | 8 - 9 | 3866 | 129 | |
NM_001335081.1 | NP_001324427.1 | 9 - 10 | 3343 | 129 | ||
NM_001335082.1 | NP_001324429.1 | 9 - 10 | 3052 | 129 | ||
NM_126244.3 | NP_565277.1 | 9 - 10 | 3369 | 129 | ||
NM_179594.2 | NP_849925.1 | 9 - 10 | 3348 | 129 | ||
NM_201667.1 | NP_973396.1 | 10 - 11 | 3069 | 129 | ||
GenBank mRNA | AB046871.1 | - | 9 - 10 | 3007 | 129 | |
AJ278528.1 | - | 9 - 10 | 3077 | 129 | ||
AJ278529.1 | - | 9 - 10 | 2960 | 129 | ||
AJ278530.1 | - | 10 - 11 | 3069 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
