Assay Details
| Gene Symbol: GAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126261.2 | NP_178309.1 | 2 - 3 | 565 | 92 | |
| GenBank mRNA | AK118125.1 | - | 2 - 3 | 565 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GAD3| Gene Symbol: GAD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126261.2 | NP_178309.1 | 2 - 3 | 565 | 92 | |
| GenBank mRNA | AK118125.1 | - | 2 - 3 | 565 | 92 | |
Set Membership: |
|