Assay Details
| Gene Symbol: GAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335087.1 | NP_001324150.1 | 1 - 2 | 590 | 63 | |
| NM_126262.2 | NP_178310.1 | 1 - 2 | 243 | 63 | ||
| GenBank mRNA | AF361836.1 | - | 1 - 2 | 187 | 63 | |
| AY124860.1 | - | 1 - 2 | 88 | 63 | ||
More Information
Set Membership: |
|

GAD4
Genomic Map