Assay Details
| Gene Symbol: PTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202569.1 | NP_001189498.1 | 3 - 4 | 396 | 135 | |
| NM_126263.3 | NP_178311.1 | 3 - 4 | 520 | 135 | ||
More Information
Set Membership: |
|

PTR4
Genomic Map