Assay Details
| Gene Symbol: PTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_126265.4 | NP_178313.1 | 3 - 4 | 1073 | 82 | |
| GenBank mRNA | AF372946.1 | - | 3 - 4 | 1048 | 82 | |
| AY143954.1 | - | 3 - 4 | 929 | 82 | ||
| L39082.1 | - | 3 - 4 | 962 | 82 | ||
More Information
Set Membership: |
|

PTR2
Genomic Map