Assay Details
| Gene Symbol: UBC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335134.1 | NP_001325163.1 | 2 - 3 | 669 | 106 | |
| NM_001335135.1 | NP_001325164.1 | 2 - 3 | 595 | 106 | ||
| NM_001335136.1 | NP_001325162.1 | 3 - 4 | 516 | 106 | ||
| NM_126331.4 | NP_565289.1 | 2 - 3 | 414 | 106 | ||
| GenBank mRNA | AF370558.1 | - | 2 - 3 | 271 | 106 | |
| AY072479.1 | - | 1 - 2 | 122 | 106 | ||
| DQ027017.1 | - | 1 - 2 | 122 | 106 | ||
| Y13031.1 | - | 247 | 106 | |||
More Information
Set Membership: |
|

UBC2
Genomic Map