Assay Details
| Gene Symbol: emb1579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335170.1 | NP_001323719.1 | 13 - 14 | 3281 | 100 | |
| NM_126366.5 | NP_178414.2 | 13 - 14 | 3323 | 100 | ||
More Information
Set Membership: |
|

emb1579
Genomic Map