Assay Details
| Gene Symbol: DOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111008.3 | NP_186791.1 | 2 - 3 | 553 | 100 | |
| GenBank mRNA | AY042787.1 | - | 2 - 3 | 485 | 100 | |
| AY064666.1 | - | 2 - 3 | 374 | 100 | ||
More Information
Set Membership: |
|

DOX1
Genomic Map