Assay Details
| Gene Symbol: DOX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111008.3 | NP_186791.1 | 9 - 10 | 1871 | 147 | |
| GenBank mRNA | AY042787.1 | - | 9 - 10 | 1803 | 147 | |
| AY064666.1 | - | 9 - 10 | 1692 | 147 | ||
More Information
Set Membership: |
|

DOX1
Genomic Map