Assay Details
| Gene Symbol: AK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111068.4 | NP_186851.1 | 4 - 5 | 1068 | 73 | |
| GenBank mRNA | AK226200.1 | - | 4 - 5 | 991 | 73 | |
| AY088366.1 | - | 4 - 5 | 990 | 73 | ||
More Information
Set Membership: |
|

AK3
Genomic Map