Assay Details
Gene Symbol: TMAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111081.3 | NP_566163.1 | 1 | 1475 | 66 | |
GenBank mRNA | AY034949.1 | - | 1 | 962 | 66 | |
AY062992.1 | - | 1 | 861 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
