Assay Details
| Gene Symbol: BIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337388.1 | NP_001327521.1 | 13 - 14 | 14623 | 80 | |
| NM_001337389.1 | NP_001327522.1 | 13 - 14 | 14551 | 80 | ||
| NM_001337390.1 | NP_001327520.1 | 13 - 14 | 14494 | 80 | ||
| NM_111093.2 | NP_186875.2 | 13 - 14 | 14495 | 80 | ||
More Information
Set Membership: |
|

BIG
Genomic Map