Assay Details
Gene Symbol: BIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337388.1 | NP_001327521.1 | 4 - 5 | 6798 | 110 | |
NM_001337389.1 | NP_001327522.1 | 4 - 5 | 6726 | 110 | ||
NM_001337390.1 | NP_001327520.1 | 4 - 5 | 6669 | 110 | ||
NM_111093.2 | NP_186875.2 | 4 - 5 | 6637 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
