Assay Details
Gene Symbol: BIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337388.1 | NP_001327521.1 | 6 - 7 | 7574 | 103 | |
NM_001337389.1 | NP_001327522.1 | 6 - 7 | 7502 | 103 | ||
NM_001337390.1 | NP_001327520.1 | 6 - 7 | 7445 | 103 | ||
NM_111093.2 | NP_186875.2 | 6 - 7 | 7434 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
