Assay Details
Gene Symbol: TRXF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111141.3 | NP_186922.1 | 1 - 2 | 399 | 99 | |
GenBank mRNA | AY065391.1 | - | 1 - 2 | 381 | 99 | |
AY084778.1 | - | 1 - 2 | 391 | 99 | ||
AY096721.1 | - | 1 - 2 | 297 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
