Assay Details
Gene Symbol: ZFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111151.5 | NP_566183.1 | 1 - 2 | 360 | 93 | |
GenBank mRNA | AK117978.1 | - | 1 - 2 | 261 | 93 | |
BT025966.1 | - | 1 - 2 | 57 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
