Assay Details
Gene Symbol: ZFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337436.1 | NP_001325699.1 | 4 - 5 | 1036 | 64 | |
NM_111151.5 | NP_566183.1 | 5 - 6 | 939 | 64 | ||
GenBank mRNA | AK117978.1 | - | 5 - 6 | 840 | 64 | |
BT025966.1 | - | 5 - 6 | 636 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
