Assay Details
| Gene Symbol: LCAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111202.4 | NP_566201.1 | 4 - 5 | 792 | 92 | |
| GenBank mRNA | AF421148.1 | - | 4 - 5 | 577 | 92 | |
| AY054239.1 | - | 4 - 5 | 610 | 92 | ||
| AY056414.1 | - | 4 - 5 | 667 | 92 | ||
| AY113174.1 | - | 4 - 5 | 577 | 92 | ||
More Information
Set Membership: |
|

LCAT3
Genomic Map