Assay Details
| Gene Symbol: LCAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111202.4 | NP_566201.1 | 9 - 10 | 1337 | 86 | |
| GenBank mRNA | AF421148.1 | - | 9 - 10 | 1122 | 86 | |
| AY054239.1 | - | 9 - 10 | 1155 | 86 | ||
| AY056414.1 | - | 9 - 10 | 1212 | 86 | ||
| AY113174.1 | - | 9 - 10 | 1122 | 86 | ||
More Information
Set Membership: |
|

LCAT3
Genomic Map