Assay Details
| Gene Symbol: DEG7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337469.1 | NP_001326670.1 | 23 - 24 | 3141 | 73 | |
| NM_111209.3 | NP_566204.2 | 22 - 23 | 3240 | 73 | ||
| GenBank mRNA | AY078951.1 | - | 22 - 23 | 3125 | 73 | |
| BT002727.1 | - | 22 - 23 | 3020 | 73 | ||
More Information
Set Membership: |
|

DEG7
Genomic Map