Assay Details
| Gene Symbol: PyrD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118214.4 | NP_567618.1 | 1 - 2 | 1203 | 73 | |
| GenBank mRNA | AK118717.1 | - | 1 - 2 | 501 | 73 | |
| BT006051.1 | - | 1 - 2 | 370 | 73 | ||
More Information
Set Membership: |
|

PyrD
Genomic Map