Assay Details
Gene Symbol: VAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341497.1 | NP_001320023.1 | 5 - 6 | 1186 | 73 | |
NM_001341498.1 | NP_001329314.1 | 5 - 6 | 1145 | 73 | ||
NM_118275.4 | NP_193886.3 | 5 - 6 | 1126 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
