Assay Details
Gene Symbol: AT4G22214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118345.4 | NP_567654.1 | 1 - 2 | 525 | 94 | |
GenBank mRNA | AK229510.1 | - | 1 - 2 | 127 | 94 | |
BT008555.1 | - | 1 - 2 | 79 | 94 | ||
BT008666.1 | - | 1 - 2 | 127 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
